Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGF2-alpha, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89137639 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-639 Supplier Abnova Corporation Supplier No. PAB13625
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against PGF2-alpha.

Specifications

Antigen PGF2-alpha
Applications Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against PGF2-alpha.
Dilution Radioimmunoassay (1:10000) The optimal working dilution should be determined by the end user.
Formulation In antiserum
Host Species Sheep
Immunogen PGF2-alpha.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity PGF2a 100% 6-keto PGF1a 0.9% Thromboxane B2 16% 13, 14-dihydro-15-keto PGF2a 0.06% 13, 14-dihydro-6.15diketo PGF2a <0.01% PGD2 0.2% PGE1 0.1% PGE2 0.3% 6-keto PGE1 0.03% PGF1a 21% Arachidonic acid <0.001%
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.