Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLG Sheep anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117346 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-346 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-346 Supplier Abnova Corporation Supplier No. PAB14917
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against PLG.

Plasminogen (PLG) is a circulating zymogen that is converted to the active enzyme plasmin by cleavage of the peptide bond between arg560 and val561, which is mediated by urokinase (PLAU; MIM 191840) and tissue plasminogen activator (PLAT; MIM 173370). The main function of plasmin is to dissolve fibrin (see, e.g., FGA, MIM 134820) clots. Plasmin, like trypsin, belongs to the family of serine proteinases (Miyata et al., 1982 [PubMed 6216475]; Forsgren et al., 1987 [PubMed 3030813]).[supplied by OMIM

Specifications

Antigen PLG
Applications ELISA, Immunocytochemistry, Immunodiffusion, Immunofluorescence, Immunomicroscopy, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against PLG.
Dilution ELISA (1:1000) Immunodiffusion (1:32) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene PLG
Gene Alias DKFZp779M0222
Gene Symbols PLG
Host Species Sheep
Immunogen Native purified human PLG.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5340
Test Specificity This antibody has single antigen reactivity with minimal cross reactivity with other human serum proteins.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.