Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Phosphatase 2 C alpha/beta, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89136967 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-967 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-967 Supplier Abnova Corporation Supplier No. PAB10355
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against synthetic peptide of Protein Phosphatase 2 C alpha/beta.

Specifications

Antigen Protein Phosphatase 2 C alpha/beta
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against synthetic peptide of Protein Phosphatase 2 C alpha/beta.
Dilution Western blot (5 to 10 ug/mL) Immunoprecipitation (5 to 10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Host Species Sheep
Immunogen A synthetic peptide (conjugated with KLH) corresponding to Protein Phosphatase 2 C alpha/beta.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.