Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ronidazole, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against Ronidazole.
Supplier: Abnova Corporation PAB8354
Specifications
Ronidazole | |
Polyclonal | |
Sheep polyclonal antibody raised against Ronidazole. | |
In 20mM phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide) | |
Ronidazole conjugated with BTG. | |
RUO | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
Unconjugated | |
Enzyme Immunoassay (10 ug/mL) The optimal working dilution should be determined by the end user. | |
Sheep | |
100 μL | |
Primary | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction