Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A1 Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89137692 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-692 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-692 Supplier Abnova Corporation Supplier No. PAB13820

Sheep polyclonal antibody raised against S100A1.

Specifications

Antigen S100A1
Applications Immunocytochemistry, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.4
Host Species Sheep
Immunogen Alum denatured porcine S100A1.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Human, Pig, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.