Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salbutamol Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89138773 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-773 Supplier Abnova Corporation Supplier No. PAB8356

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Sheep polyclonal antibody raised against Salbutamol.

Specifications

Antigen Salbutamol
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against Salbutamol.
Dilution Enzyme Immunoassay (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide)
Host Species Sheep
Immunogen Salbutamol conjugated with HSA.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Carbuterol 90%, Salbutamol 86%, Methyl-Clenbuterol 75%, Brombuterol 53%, Terbutaline 50%, Mabuterol 45%, Pirbuterol 25%, Mapenterol 32%
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.