Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINF1/SERPINF2, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89117349 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-349 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-349 Supplier Abnova Corporation Supplier No. PAB14922
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against SERPINF1/SERPINF2.

Specifications

Antigen SERPINF1/SERPINF2
Applications ELISA, Immunocytochemistry, Immunodiffusion, Immunofluorescence, Immunomicroscopy, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against SERPINF1/SERPINF2.
Dilution ELISA (1:1000) Immunodiffusion (1:32) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene Alias EPC-1/PEDF/PIG35
Gene Symbols SERPINF1
Host Species Sheep
Immunogen Purified human SERPINF1/SERPINF2.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5176/5345
Test Specificity This antibody has a single antigen reactivity with minimal cross reactivity with other human serum proteins.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.