Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNCB Sheep anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114409 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-409 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-409 Supplier Abnova Corporation Supplier No. PAB0769
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against synthetic peptide of SNCB.

Sequence: IEPLMEPEGSYEDPPQE

Specifications

Antigen SNCB
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against synthetic peptide of SNCB.
Dilution Immunohistochemistry (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene SNCB
Gene Symbols SNCB
Host Species Sheep
Immunogen A synthetic peptide (conjugated with diphteria toxid) corresponding to amino acids 108-125 of human SNCB.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6620
Test Specificity Known to be specific to beta synuclein.
Target Species Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.