Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSeCKS, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89136968 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-968 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-968 Supplier Abnova Corporation Supplier No. PAB10358
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against partial recombinant SSeCKS.

Specifications

Antigen SSeCKS
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against partial recombinant SSeCKS.
Dilution Western blot (2 to 5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Host Species Sheep
Immunogen Recombinant protein corresponding to amino acids 620-1596 of SSeCKS.
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.