Learn More
SUMO1, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against synthetic peptide of SUMO1.
Supplier: Abnova Corporation PAB0765
Description
This gene encodes a protein that is a member of the SUMO (small ubiquitin-like modifier) protein family. It functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. However, unlike ubiquitin which targets proteins for degradation, this protein is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability. It is not active until the last four amino acids of the carboxy-terminus have been cleaved off. Several pseudogenes have been reported for this gene. Alternate transcriptional splice variants encoding different isoforms have been characterized. [provided by RefSeq
Sequence: AKPSTEDLGDKKEGEYSpecifications
SUMO1 | |
Polyclonal | |
Sheep polyclonal antibody raised against synthetic peptide of SUMO1. | |
Lyophilized from PBS | |
DAP-1/GMP1/OFC10/PIC1/SENP2/SMT3/SMT3C/SMT3H3/SUMO-1/UBL1 | |
Sheep | |
100 μL | |
Primary | |
This antiserum recognizes human SUMO-1 and not ubiquitin. | |
Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (Frozen), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:2000-1:4000) Western Blot (1:4000-1:8000) The optimal dilution should be determined by the end user. | |
SUMO1 | |
SUMO1 | |
A synthetic peptide (conjugated diphtheria toxoid) corresponding to amino acids 6-21 of human SUMO1. | |
RUO | |
7341 | |
Human, Rat | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only