Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THPO, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89116853 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-853 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-853 Supplier Abnova Corporation Supplier No. PAB13838

Sheep polyclonal antibody raised against recombinant Thpo.

Specifications

Antigen THPO
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description thrombopoietin
Formulation In 50mM sodium phosphate buffer, 100mM NaCl, 1mM EDTA, pH 6.6
Gene Thpo
Gene Alias Mpllg, TPO, TPO-1, TPO-2, TPO-3, TPO-4
Gene Symbols Thpo
Host Species Sheep
Immunogen Recombinant protein corresponding to mouse Thpo.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21832
Test Specificity Recognizes Mouse Thrombopoietin.
Target Species Mouse
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.