Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thromboxane B2, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89137653 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-653 Supplier Abnova Corporation Supplier No. PAB13667
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against Thromboxane B2.

Specifications

Antigen Thromboxane B2
Applications ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against Thromboxane B2.
Dilution Radioimmunoassay (1:10000) The optimal working dilution should be determined by the end user.
Host Species Sheep
Immunogen Thromboxane B2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This is a sheep polyclonal antibody specific to Thromboxane B2 (TXB2). HAPTEN REACTIVITY: TXB2 100% 6,keto-PGF1alpha 0.17% PGF1alpha 0.14% PGF2alpha 0.13% PGD2 <0.07% 6-KetoPGF1alpha <0.07% 13,14-dihydro-15-keto PGF2alpha <0.07% 13,14-dihydro-6,15diket PGF2alpha <0.07% PGE1 <0.07% PGE3 <0.07% Arachidonic Acid <0.07%
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.