Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSX2 Sheep anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against partial recombinant VSX2.
Supplier: Abnova Corporation PAB9842
Specifications
VSX2 | |
Polyclonal | |
Sheep polyclonal antibody raised against partial recombinant VSX2. | |
In PBS (0.08% sodium azide) | |
CHX10/HOX10/MCOP2/MCOPCB3/RET1 | |
Sheep | |
250 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
Unconjugated | |
Western Blot (0.5-1.0 ug/mL) The optimal working dilution should be determined by the end user. | |
VSX2 | |
VSX2 | |
Recombinant GST fusion protein corresponding to amino acids 264-361 of human VSX2. | |
RUO | |
338917 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction