Learn More
Carolina™ Formalin Preserved Sheep Brain, Dura Mater Removed
SDPSCI_ED
Catalog No. s28069
Click to view available options
Packaging:
Bag
Pail
Carolina™ Formalin Preserved Sheep Brain with the Dura Mater Removed is used to study mammalian brain structure.
  • Similar to human brain
  • Sheep brain with dura mater removed and optic chiasma intact are shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Odorless fluid designed to minimize unpleasant odor of formaldehyde
  • Plain: No color injection
  • Individually damp-packed in vacuum-sealed bags
Specifications
Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 1/Pk.
Packaging Bag
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.