Learn More
Description
Specifications
Specifications
| Antigen | IgG |
| Conjugate | Alkaline Phosphatase (AP) |
| Formulation | 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Gene Symbols | IGHG1 |
| Host Species | Sheep |
| Purification Method | Multi-step |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Primary or Secondary | Isotype Control (Primary) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
