Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Sheep Triiodothyronine ELISA Kit (Colorimetric)
SDP

Catalog No. NB601771KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB601771KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB601771KT Supplier Novus Biologicals™ Supplier No. NBP2601771KIT
Add to Cart
Add to Cart

Assay has high sensitivity and specificity for detection of sheep T3.

The Sheep Triiodothyronine ELISA Kit (Colorimetric) measures sheep Triiodothyronine in serum, plasma, cell culture supernates, tissue homogenates.

Specifications

Assay Range 0.5ng/mL to 8ng/mL
Assay Sensitivity 0.5ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates
Specificity For detection of sheep T3
Target Species Sheep
For Use With (Application) ELISA
Kit Contents Assay plate, Adhesive Strip (For 96 wells), HRP-avidin, Conjugate, Substrate B, Substrate A, Wash Buffer (20 x concentrate), Stop Solution, Standard, Instruction manual
Quantity 1 Kit
Research Discipline Signal Transduction
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym liothyronine; T3
Target Triiodothyronine
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.