Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566081 Supplier Invitrogen™ Supplier No. PA566081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPGHRKRASPK GSLSSAQPHS WMLTPSPLNS HCAHREPISS SPQPVANGPK QKKKSNWRST TRLRIIRLRD Highest antigen sequence identity to the following orthologs - mouse 31%, rat 32%.

Adapter protein which may play a role in the regulation of apoptosis in response to PDGF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHF
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SHF
Gene Accession No. Q7M4L6
Gene Alias CTD-2651B20.7; RGD1562705; SH2 domain-containing adapter protein F; SH2 domain-containing adapter protein F; LOW QUALITY PROTEIN: SH2 domain-containing adapter protein F; shf; Src homology 2 domain containing F
Gene Symbols SHF
Host Species Rabbit
Immunogen Recombinant Human SHF. Recombinant protein control fragment (Product #RP-106772).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.