Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SHFM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567253
Description
Immunogen sequence: MSEKKQPVDLG LLEEDDEFEE FPAEDWAGLD ED Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The product of this gene has been localized within the split hand/split foot malformation locus SHFM1 at chromosome 7. It has been proposed to be a candidate gene for the autosomal dominant form of the heterogeneous limb developmental disorder split hand/split foot malformation type 1. In addition, it has been shown to directly interact with BRCA2. It also may play a role in the completion of the cell cycle.
Specifications
SHFM1 | |
Polyclonal | |
Unconjugated | |
SEM1 | |
26S proteasome complex subunit DSS1; 26S proteasome complex subunit SEM1; C7orf76; Deleted in split hand/split foot protein 1; deleted in split hand/split foot protein 1 homolog; deleted in split-hand/split-foot 1; Dss1; ECD; Putative protein SEM1, isoform 2; Sem1; SHFD1; Shfdg1; Shfg; Shfm1; SHSF1; split hand/foot deleted gene 1; split hand/foot deleted protein 1; Split hand/foot deleted protein 1 homolog; split hand/foot malformation (ectrodactyly) type 1; split hand/foot malformation type 1 protein; Split hand/foot malformation type 1 protein homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
0, 7979 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P60896, Q6ZVN7 | |
SEM1 | |
Recombinant Human SHFM1. Recombinant protein control fragment (Product #RP-107889). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction