Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEM1 isoform 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115241
Description
Antibody detects endogenous levels of total FLJ42280.
SEM1 (SEM1 26S Proteasome Subunit) is a Protein Coding gene. The product of this gene has been localized within the split hand/split foot malformation locus SHFM1 at chromosome 7. It has been proposed to be a candidate gene for the autosomal dominant form of the heterogeneous limb developmental disorder split hand/split foot malformation type 1. In addition, it has been shown to directly interact with BRCA2. It also may play a role in the completion of the cell cycle.
Specifications
| SEM1 isoform 2 | |
| Polyclonal | |
| Unconjugated | |
| SEM1 | |
| 26S proteasome complex subunit DSS1; 26S proteasome complex subunit SEM1; C7orf76; Deleted in split hand/split foot protein 1; deleted in split hand/split foot protein 1 homolog; deleted in split-hand/split-foot 1; Dss1; ECD; Putative protein SEM1, isoform 2; Sem1; SHFD1; Shfdg1; Shfg; Shfm1; SHSF1; split hand/foot deleted gene 1; split hand/foot deleted protein 1; Split hand/foot deleted protein 1 homolog; split hand/foot malformation (ectrodactyly) type 1; split hand/foot malformation type 1 protein; Split hand/foot malformation type 1 protein homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 0, 680532, 7979 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P60896, Q6ZVN7 | |
| SEM1 | |
| A synthesized peptide derived from human SEM1(Accession Q6ZVN7), corresponding to amino acid residues S91-C128. | |
| 100 μL | |
| Primary | |
| Human, Rat, Monkey | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction