Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHFM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554204
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554204 Supplier Invitrogen™ Supplier No. PA554204
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CQDSNICAVF AVQGGKVGRK HGIKRGRRPS IRSPAQRARG PWIHESKHPA FAKQQINLEM P Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.

The product of this gene has been localized within the split hand/split foot malformation locus SHFM1 at chromosome 7. It has been proposed to be a candidate gene for the autosomal dominant form of the heterogeneous limb developmental disorder split hand/split foot malformation type 1. In addition, it has been shown to directly interact with BRCA2. It also may play a role in the completion of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHFM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEM1
Gene Accession No. Q6ZVN7
Gene Alias 26S proteasome complex subunit DSS1; 26S proteasome complex subunit SEM1; C7orf76; Deleted in split hand/split foot protein 1; deleted in split hand/split foot protein 1 homolog; deleted in split-hand/split-foot 1; Dss1; ECD; Putative protein SEM1, isoform 2; Sem1; SHFD1; Shfdg1; Shfg; Shfm1; SHSF1; split hand/foot deleted gene 1; split hand/foot deleted protein 1; Split hand/foot deleted protein 1 homolog; split hand/foot malformation (ectrodactyly) type 1; split hand/foot malformation type 1 protein; Split hand/foot malformation type 1 protein homolog
Gene Symbols SEM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SHFM1. Recombinant protein control fragment (Product #RP-92848).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.