Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHH Recombinant Rabbit Monoclonal Antibody (HL2076)
GREENER_CHOICE

Catalog No. PIMA547279
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547279 Supplier Invitrogen™ Supplier No. MA547279
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Sonic Hedgehog (SHH), which is expressed only during embryogenesis, is instrumental in patterning the early embryo. It has been implicated as the key inductive signal in patterning of the ventral neural tube, the anterior-posterior limb axis, and the ventral somites. Of three human proteins showing sequence and functional similarity to the Sonic Hedgehog protein of Drosophila, this protein is the most similar. The protein is made as a precursor that is autocatalytically cleaved, the N-terminal portion is soluble and contains the signalling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the developing embryo. Defects in this protein or in its signalling pathway are a cause of holoprosencephaly (HPE), a disorder in which the developing forebrain fails to correctly separate into right and left hemispheres. HPE is manifested by facial deformities. In addition, it is thought that mutations in this gene or in its signalling pathway may be responsible for VACTERL syndrome, which is characterized by vertebral defects, anal atresia, tracheoesophageal fistula with esophageal atresia, radial and renal dysplasia, cardiac anomalies, and limb abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHH
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL2076
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Shh
Gene Accession No. Q15465, Q62226, Q63673
Gene Alias 9530036O11Rik; Dsh; fc83d08; hedgehog; hemimelic extra toes; Hhg1; HHG-1; HLP3; HPE3; Hx; Hxl3; M100081; MCOPCB5; morphogen; shh; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; shh.L; shha; ShhN; ShhNC; ShhNp; short digits; SMMCI; sonic hedgehog; sonic hedgehog a; sonic hedgehog homolog; sonic hedgehog L homeolog; sonic hedgehog protein; Sonic hedgehog protein 19 kDa product; Sonic hedgehog protein A; Sonic hedgehog protein A N-product; Sonic hedgehog protein N-product; sonic hedgehog signaling molecule; sonic hedgehog signaling molecule a; sonic-you; syu; TPT; TPTPS; vhh1; vhh-1; wu:fc83d08; XELAEV_18030676mg; Xhh; xshh; X-SHH
Gene Symbols Shh
Host Species Rabbit
Immunogen Synthetic peptide of human Shh.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20423, 29499, 6469
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.