Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shh Mouse anti-Mouse, Clone: 67.8, Abnova™

Catalog No. 89035562 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-562 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-562 Supplier Abnova Corporation Supplier No. MAB14164

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant mouse Shh.

  • High specificity
  • High reactivity

Specifications

Antigen Shh
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 67.8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant mouse Shh.
Dilution Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide).
Gene Accession No. Q62226
Gene Alias 9530036O11Rik/Dsh/Hhg1/Hx/Hxl3/M100081
Gene Symbols Shh
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminus of mouse Shh.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20423
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.