Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588595 Supplier Invitrogen™ Supplier No. PA588595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549; Cellular Location: Cell membrane, Lipid-anchor, extracellular space Immunogen sequence: GRLLYSDFLT FLDRDDGAKK VFYVIETREP RERLLLTAAH LLFVAPHNDS ATGEPEASSG SGPPSGGALG PRALFASRVR PGQRVYVVAE RDGDRRLLPA.

Sonic Hedgehog (SHH), which is expressed only during embryogenesis, is instrumental in patterning the early embryo. It has been implicated as the key inductive signal in patterning of the ventral neural tube, the anterior-posterior limb axis, and the ventral somites. Of three human proteins showing sequence and functional similarity to the Sonic Hedgehog protein of Drosophila, this protein is the most similar. The protein is made as a precursor that is autocatalytically cleaved, the N-terminal portion is soluble and contains the signalling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the developing embryo. Defects in this protein or in its signalling pathway are a cause of holoprosencephaly (HPE), a disorder in which the developing forebrain fails to correctly separate into right and left hemispheres. HPE is manifested by facial deformities. In addition, it is thought that mutations in this gene or in its signalling pathway may be responsible for VACTERL syndrome, which is characterized by vertebral defects, anal atresia, tracheoesophageal fistula with esophageal atresia, radial and renal dysplasia, cardiac anomalies, and limb abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHH
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Shh
Gene Accession No. Q15465
Gene Alias 9530036O11Rik; Dsh; fc83d08; hedgehog; hemimelic extra toes; Hhg1; HHG-1; HLP3; HPE3; Hx; Hxl3; M100081; MCOPCB5; morphogen; shh; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; shh.L; shha; ShhN; ShhNC; ShhNp; short digits; SMMCI; sonic hedgehog; sonic hedgehog a; sonic hedgehog homolog; sonic hedgehog L homeolog; sonic hedgehog protein; Sonic hedgehog protein 19 kDa product; Sonic hedgehog protein A; Sonic hedgehog protein A N-product; Sonic hedgehog protein N-product; sonic hedgehog signaling molecule; sonic hedgehog signaling molecule a; sonic-you; syu; TPT; TPTPS; vhh1; vhh-1; wu:fc83d08; XELAEV_18030676mg; Xhh; xshh; X-SHH
Gene Symbols Shh
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 231-330 of human Sonic Hedgehog (Shh) (NP_0001841).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6469
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.