Learn More
Invitrogen™ Shiga toxin 1B Monoclonal Antibody (A42L)

Description
This antibody is specific for Shiga toxin type I, B subunit (Stx-1b). Purity is >90%.
Specifications
Specifications
| Antigen | Shiga toxin 1B |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | A42L |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Shiga toxin; Shigatoxin Type 1; Shigatoxin Type I; STX1; STX-1b; STX-Ib |
| Host Species | Mouse |
| Immunogen | Verotoxin (shigatoxin) native protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.