Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Shiga toxin 1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119982
This item is not returnable.
View return policy
Description
Positive Control: Full length recombinant protein of shiga toxin subunit B Product has been tested in direct ELISA.

Specifications
Shiga toxin 1B | |
Polyclonal | |
Unconjugated | |
Shiga toxin; Shigatoxin Type 1; Shigatoxin Type I; STX1; STX-1b; STX-Ib | |
Full length protein of shiga toxin subunit B. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction