Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Shiga toxin 2A Monoclonal Antibody (B19J)
GREENER_CHOICE

Catalog No. MA17101
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17101 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17101 Supplier Invitrogen™ Supplier No. MA17101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7101 detects the Verotoxin. This antibody is specific for Verotoxin II - A subunit (SLT-2a). MA1-7101 has been successfully used in ELISA procedures. The MA1-7101 immunogen is purified native toxoid.

Shiga-like toxins (SLTs), are also called Verotoxins. Enterohemorrhagic Escherichia coli (EHEC) strains which are primarily of serotypes 0157:H7, 026:H11, and O111:H8 have been incriminated as etiologic agents of hemorrhagic colitis and Hemolytic-uremic syndrome, a generalized disease characterized by acute renal failure, thrombocytopenia, and microangiopathic hemolytic anemia. There are several distinct E.coli SLTs. SLT-I and SLT-II are produced by EHEC. SLT-I and Shiga toxin share 99% deduced amino acid sequence homology, whereas SLT-I and SLT-II share about 60% deduced amino acid sequence homology. SLT-I and SLT-II are antigenically distinct. Antibodies to SLT-II can also neutralize a variant of SLT-II (designated SLT-Iiv) produced by strains of E.coli that cause edema. SLT-IIv is cytotoxic for Vero but not HeLa cells, distinguishing it from SLT-II. The protein structure of the toxin consists of two domains: the A polypeptide that inhibits protein synthesis by targeting ribosomes, and the B polypeptide pentamer that binds to the eukaryotic cell receptor globotriaosylceramide (Gb3) leading to receptor-mediated endocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Shiga toxin 2A
Applications ELISA
Classification Monoclonal
Clone B19J
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Shiga toxin; Shigatoxin Type 2; Shigatoxin Type II; STX2; STX-2a; STX-IIa
Host Species Mouse
Immunogen Purified native toxoid.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.