Learn More
Invitrogen™ Shiga toxin 2B Monoclonal Antibody (7588)
Description
Specificity: Specific for Shiga toxin type II - B subunit (Stx-2b). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Specifications
Specifications
| Antigen | Shiga toxin 2B |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 7588 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Shiga toxin; Shigatoxin Type 2; Shigatoxin Type II; STX2; STX-2b; STX-IIb |
| Host Species | Mouse |
| Immunogen | Proprietary sequence. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.