Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Shigella Antibody (306/305) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062918
Description
Shigella Monoclonal specifically detects Shigella in Bacteria samples. It is validated for ELISA.Specifications
Shigella | |
Monoclonal | |
1.0 mg/mL | |
ELISA 1:100-1:2000 | |
Mouse | |
Affinity Purified | |
RUO | |
This product is specific for Shigella flexneri serotype 6. S. flexneri is a gram negative, rod shaped bacteria that is very closely related to E. coli and Salmonella. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ELISA | |
306/305 | |
Unconjugated | |
Shigella Boydii; Shigella Dysenteriae; Shigella Flexneri; Shigella Sonnei | |
NCTC 9729 | |
0.1 mg | |
Primary | |
Bacteria | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction