Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Shigella Antibody (306/305), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062918V
Description
Shigella Monoclonal specifically detects Shigella in Bacteria samples. It is validated for ELISA.Specifications
Shigella | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
This product is specific for Shigella flexneri serotype 6. S. flexneri is a gram negative, rod shaped bacteria that is very closely related to E. coli and Salmonella. | |
Store at 4C in the dark. | |
IgG1 |
ELISA | |
306/305 | |
ELISA | |
Shigella Boydii; Shigella Dysenteriae; Shigella Flexneri; Shigella Sonnei | |
NCTC 9729 | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction