Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Shigella Dysenteriae Serotype 1 Antibody (35/383), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064568AF647
Description
Shigella Dysenteriae Serotype 1 Monoclonal specifically detects Shigella Dysenteriae Serotype 1 in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Shigella Dysenteriae Serotype 1 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Reacts with NCTC 4837. Shigella Dysenteriae Serotype 1 | |
Bacteria | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
35/383 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
S. boydii, S. dysenteriae, S. flexneri, S. sonnei, Shigella Boydii, Shigella Dysenteriae, Shigella Flexneri, Shigella Sonnei | |
Native S. dysenteriae, NCTC 4837 (ATCC 13313). | |
0.1 mL | |
Primary | |
Protein A purified | |
Store at 4C in the dark. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction