Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Shigella Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA185871
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA185871 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA185871 Supplier Invitrogen™ Supplier No. PA185871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is >95%. Rabbit anti Shigella group antigen antibody is polyvalent for Shigella antigens. Shigella sp. are non spore forming, rod shaped bacteria, the causative agent of Shigellosis in man. The genus is closely related to Escherichia coli and Salmonella. Rabbit anti Shigella group antigen antibody preparation has not been cross absorbed and is likely to react with closely related Enterobacteriaceae.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Shigella
Applications ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Host Species Rabbit
Immunogen Mixture of S. boydii, S. flexneri and S. dysenteriae.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.