Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Shigella Polyclonal Antibody, FITC
GREENER_CHOICE

Catalog No. PIPA173026
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173026 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173026 Supplier Invitrogen™ Supplier No. PA173026
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73026 antibody reacts with Shigella Group antigen. PA1-73026 has been successfully used in immunofluorescence applications. The PA1-73026 antibody was raised against a mixture of S. boydii, S. flexneri & S. dysenteriae.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Shigella
Applications Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Host Species Rabbit
Immunogen Mixture of S. boydii, S. flexneri & S. dysenteriae.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.