Learn More
Invitrogen™ Shigella sonnei Form 1 LPS Monoclonal Antibody (0946)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547614
Description
Specificity: Specific to the LPS of Shigella sonnei form 1. No cross reactivity with: S. sonnei form 2, S. flexneri (mix), S. dysenteriae, S. boydii, E. coli (mix), Salmonellae (mix) and Citrobacter. IS cross reactive with Plesiomonas shigelloides, as expected. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Shigella sonnei is a rod-shaped, gram-negative facultative intracellular pathogen. Together with Shigella flexneri, it is responsible for the majority of shigellosis cases.
Specifications
Shigella sonnei Form 1 LPS | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2a |
ELISA, Immunocytochemistry | |
0946 | |
Unconjugated | |
S. sonnei | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.