Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Shikimic acid, 98%

Catalog No. aal04848md
Click to view available options
Quantity:
250 mg
1 g

CAS: 138-59-0 | C7H9O5- | 173.144 g/mol

Antagonistic antimicrobial agent which also inhibits rapamycin biosynthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-59-0
Molecular Formula C7H9O5-
Molecular Weight (g/mol) 173.144
MDL Number MFCD00066278
InChI Key JXOHGGNKMLTUBP-HSUXUTPPSA-M
Synonym shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
PubChem CID 7057976
ChEBI CHEBI:36208
IUPAC Name (3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
SMILES C1C(C(C(C=C1C(=O)[O-])O)O)O

Specifications

Melting Point 184°C to 188°C
Odor Odorless
Quantity 250 mg
Beilstein 4782717
Sensitivity Hygroscopic
Merck Index 14,8480
Optical Rotation −175° (c=2 in Water)
Formula Weight 174.16
Percent Purity 98%
Chemical Name or Material (-)-Shikimic acid
EINECSNumber 205-334-2
RTECSNumber GW4600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.