Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shikimic Acid, MP Biomedicals™

Catalog No. p-7083116
Click to view available options
Quantity:
100 mg
500 mg

More commonly known as its anionic form shikimate. Shikimic Acid, MP Biomedicals is a biosynthetic precursor of aromatic amino acids and many alkaloids. It is an important biochemical metabolite in plants and microorganisms and is important in the biosynt

Shikimic acid is a precursor for the aromatic amino acids (phenylalanine, tyrosine and trypyophan), indole and indole derivatives. It is also a precursor of many alkaloids and other aromatic metabolites, phenylpropanoids, flavonoids, tannins, and lignin.

This tri-hydroxy cyclohexene carboxylic acid is important in the biosynthesis of so many compounds that the shikimate pathway (the seven-step metabolic route used by bacteria, fungi, algae, parasites and plants for the biosynthesis of aromatic amino acids) is named after it.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-59-0
Molecular Formula C7H9O5-
Molecular Weight (g/mol) 173.144
InChI Key JXOHGGNKMLTUBP-HSUXUTPPSA-M
Synonym shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
PubChem CID 7057976
ChEBI CHEBI:36208
IUPAC Name (3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
SMILES C1C(C(C(C=C1C(=O)[O-])O)O)O

Specifications

Melting Point 182°C to 192°C
Quantity 100 mg
Solubility Information Solubility (50mg/mL H2O): Clear to Slightly Hazy
Optical Rotation −185 to −181° (C=4 in H2O at 20°C) (Lit.)
Formula Weight 174.2
Percent Purity ≥98%
Physical Form Powder
Recommended Storage Room Temperature (15–30°C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.