Learn More
Restek™ Shimadzu HPLC Systems: Seals
SDP
Catalog No. 06712839 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Model:
LC-10 Ai, 10 AS, 10 AT, 10 ATvp, LC-7 A
LC-10 Ai, LC-10 AS, LC-10 AT, LC-10 ATvp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
06-712-839 LC-10 Ai, LC-10 AS, LC-10 AT, LC-10 ATvp
06-712-837 LC-10 Ai, 10 AS, 10 AT, 10 ATvp, LC-7 A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-712-839 Supplier Restek™ Supplier No. 25292
For use with Shimadzu HPLC Systems.
Plunger Seal, Gold
  • Ultra-high molecular weight polyethylene (UHMWPE)
  • Increased resistance to abrasion
  • Good for buffers
Specifications
Type Seal
Model LC-10 Ai, LC-10 AS, LC-10 AT, LC-10 ATvp
For Use With Shimadzu HPLC Systems