Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHIP Antibody (SHIP-01), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11081538AF750
Description
SHIP Monoclonal specifically detects SHIP in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
SHIP | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
INPP5D | |
Peptide coresponding to a sequence within N-terminal domain of Human SHIP-1. | |
Primary | |
The SHIP-01 reacts with SHIP-1, a phosphoinositide phosphatase largely confined to hematopoietic cells. Multiple forms of SHIP-1 have been reported with molecular weights of 110, 125, 130, 135 and 145 kDa. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
SHIP-01 | |
Western Blot, Flow Cytometry | |
145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140 | |
Mouse | |
0.1 mL | |
3635 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction