Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHIP Antibody (SHIP-02) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP145077
Description
SHIP Monoclonal specifically detects SHIP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
SHIP | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry, Flow (Intracellular) 2-5 ug/ml, CyTOF-ready | |
INPP5D | |
Peptide coresponding to a sequence within N-terminal domain of Human SHIP-1. | |
Protein A purified | |
RUO | |
Primary | |
The antibody SHIP-02 reacts with SHIP-1, a phosphoinositide phosphatase largely confined to hematopoietic cells. Multiple forms of SHIP-1 have been reported with molecular weights of 110, 125, 130, 135 and 145 kDa. | |
Store at 4C. Do not freeze. | |
IgG2a |
Western Blot, Flow Cytometry, CyTOF | |
SHIP-02 | |
Unconjugated | |
145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140 | |
Mouse | |
145 kDa | |
0.1 mg | |
Adaptive Immunity, Immunology, Signal Transduction | |
3635 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction