Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHIP, p Tyr1020 Antibody (SR1167), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321565100UL
Description
SHIP Monoclonal antibody specifically detects SHIP in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
SHIP | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
SR1167 | |
Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:20 | |
145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140 | |
A synthesized peptide derived from human phospho-SHIP [Tyr1020] (Uniprot #: Q92835) | |
100 μg | |
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
3635 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction