Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587643 Supplier Invitrogen™ Supplier No. PA587643
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLYAQPNQGL VCALLLPVEG EREPDPPDDR DASDGEDEKP PLPPRSGSTS ISAPTGPSSP LPAPETPTAP AAESAPNGLS TVSHDYLKGS YGLDLEAVRG GASHLPHLTR TLATSCRRLH SEVDKVLSGL EILSKVFDQQ SSPMVTRLLQ QQNLPQTGEQ ELESLVLKLS VLKDFLSGIQ KKALKALQDM SSTAPPAPQP STRKAKTIPV Q; Positive Samples: HeLa, U-251MG, A-549, BT-474, Mouse heart, Mouse brain, Rat brain; Cellular Location: Cell projection, Cytoplasm, Membrane, Peripheral membrane protein, cytoskeleton, cytosol, filopodium, lamellipodium.

The protein encoded by this gene is an SH2-containing 5'-inositol phosphatase that is involved in the regulation of insulin function. The encoded protein also plays a role in the regulation of epidermal growth factor receptor turnover and actin remodeling. Additionally, this gene supports metastatic growth in breast cancer and is a valuable biomarker for breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHIP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene INPPL1
Gene Accession No. O15357, Q6P549, Q9WVR3
Gene Alias 51C; 51C protein; ablSH3-binding protein; inositol polyphosphate phosphatase like 1; inositol polyphosphate phosphatase-like 1; inositol polyphosphate phosphatase-like protein 1; Inppl1; INPPL-1; NS1; OPSMD; Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 2; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 2; protein 51C; PTP2C; SH2 domain-containing inositol 5'-phosphatase 2; SH2 domain-containing inositol phosphatase 2; SH2 domain-containing inositol-5'-phosphatase 2; SH2-containing inositol phosphatase 2; SHIP2; SHIP-2; SHP-2; SH-PTP2
Gene Symbols INPPL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-310 of human INPPL1 (NP_0015583).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16332, 3636, 65038
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.