Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHISA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565598
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565598 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565598 Supplier Invitrogen™ Supplier No. PA565598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RCLRPKQDPQQ SRAPGGNRLM ETIPMIPSAS TSRGSSSRQ Highest antigen sequence identity to the following orthologs - mouse 98%, rat 100%.

Plays an essential role in the maturation of presomitic mesoderm cells by individual attenuation of both FGF and WNT signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHISA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SHISA2
Gene Accession No. Q6UWI4
Gene Alias 9430059P22Rik; bA398O19.2; C13orf13; hShisa; MAd2; mShisa; PRO28631; protein shisa-2 homolog; shisa; shisa family member 2; shisa homolog 2; SHISA2; shisa-like protein 2; TMEM46; Transmembrane protein 46; UNQ9166/PRO28631; WGAR9166
Gene Symbols SHISA2
Host Species Rabbit
Immunogen Recombinant Human SHISA2. Recombinant protein control fragment (Product #RP-104891).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387914
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.