Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHISA3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA563016
Description
Immunogen sequence: SNNIAIKGVA RFYRSRKKHL ITTQTEHKCV LDSCRSLEAE GFQVTYLPVQ KSGIIDLKEL EAAIQPDTSL VSVMTVNNEI GVKQPIAEIG RICSSRKV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
SHISA3 plays an essential role in the maturation of presomitic mesoderm cells by individual attenuation of both FGF and WNT signaling. The Shisa family of single-transmembrane proteins is characterized by an N-terminal cysteine-rich domain and a proline-rich C-terminal region. Its founding member, Xenopus Shisa, promotes head development by antagonizing Wnt and FGF signaling. Shisa physically interacted with immature forms of the Wnt receptor Frizzled and the FGF receptor within the ER and inhibited their posttranslational maturation and trafficking to the cell surface. Loss of Shisa function sensitized the neuroectoderm to Wnt signaling and suppressed head formation during gastrulation.
Specifications
SHISA3 | |
Polyclonal | |
Unconjugated | |
SHISA3 | |
SHISA3, Shisa homolog 3, hShisa3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
152573 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A0PJX4 | |
SHISA3 | |
Recombit protein corresponding to Human SHISA3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction