Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHISA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534528
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534528 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534528 Supplier Invitrogen™ Supplier No. PA534528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-34528 can be used with blocking peptide PEP-1571.

SHISA4 plays an essential role in the maturation of presomitic mesoderm cells by individual attenuation of both FGF and WNT signaling. The Shisa family of single-transmembrane proteins is characterized by an N-terminal cysteine-rich domain and a proline-rich C-terminal region. Its founding member, Xenopus Shisa, promotes head development by antagonizing Wnt and FGF signaling. Shisa physically interacted with immature forms of the Wnt receptor Frizzled and the FGF receptor within the ER and inhibited their posttranslational maturation and trafficking to the cell surface. Loss of Shisa function sensitized the neuroectoderm to Wnt signaling and suppressed head formation during gastrulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHISA4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SHISA4
Gene Accession No. Q8CA71, Q96DD7
Gene Alias 9330132O05Rik; C1orf40; mShisa4; Protein shisa-4; shisa family member 4; shisa homolog 4; SHISA4; Tmem58; transmembrane protein 58; UNQ583/PRO1153
Gene Symbols SHISA4
Host Species Rabbit
Immunogen A 15 amino acid peptide near the center of human SHISA4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149345, 77552
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.