Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHISA5 Monoclonal Antibody (3G5)
GREENER_CHOICE

Catalog No. PIMA521886
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521886 Supplier Invitrogen™ Supplier No. MA521886
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MGFGATLAVG LTIFVLSVVT IIICFTCSCC CLYKTCRRPR PVVTTTTSTT VVHAPYPQPP SVPPSYPGPS YQGYHTMPPQ PGMPAAPYPM QYPPPYPAQP MGPPAYHETL AGGAAAPYPA SQPPYNPAYM DAPKAAL.

Can induce apoptosis in a caspase-dependent manner and plays a role in p53/TP53-dependent apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHISA5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G5
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SHISA5
Gene Accession No. Q8N114
Gene Alias NBP; NBP1; Protein shisa-5; PSEC0133; Putative NF-kappa-B-activating protein 120; SCOTIN; shisa family member 5; shisa homolog 5; SHISA5
Gene Symbols SHISA5
Host Species Mouse
Immunogen SHISA5 (AAH01463, 1 a.a. ~ 137 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51246
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.