Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHISA9 Rabbit anti-Human, Mouse, Non-human Primate, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc CKAMP44A101AP

Description
SHISA9 Polyclonal antibody specifically detects SHISA9 in Human, Mouse, Non-human Primate samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
SHISA9 | |
Polyclonal | |
Unconjugated | |
SHISA9 | |
2700045P11Rik; Ckamp44; cystine-knot AMPAR modulating protein; cystine-knot AMPAR modulating protein of 44 kDa; Protein shisa-9; shisa family member 9; shisa homolog 9; shisa homolog 9-like; SHISA9 | |
Synthetic peptide taken within amino acid region 250-300 on CKAMP44a protein. | |
200 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
B4DS77, Q9CZN4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
72555, 729993, 739962 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction