Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHKBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559773 Supplier Invitrogen™ Supplier No. PA559773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVFPVKRRNR HSLVGPQQLG GRPAPVRRSN TMPPNLGNAG LLGRMLDEKT PPSPSGQPEE PGMVRLVCGH HNWIAVAYTQ FLVCYRLKEA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

Inhibits CBL-SH3KBP1 complex mediated down-regulation of EGFR signaling by sequestration of SH3KBP1. Binds to SH3KBP1 and prevents its interaction with CBL and inhibits translocation of SH3KBP1 to EGFR containing vesicles upon EGF stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHKBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SHKBP1
Gene Accession No. Q8TBC3
Gene Alias B930062H15Rik; PP203; RGD1309281; Sb1; SETA binding protein 1; SETA-binding protein 1; SH3KBP1 binding protein 1; SH3KBP1-binding protein 1; Shkbp1
Gene Symbols SHKBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SHKBP1. Recombinant protein control fragment (Product #RP-99113).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.