Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHMT1 Antibody (AT26E5), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259437IR
This item is not returnable.
View return policy
Description
SHMT1 Monoclonal antibody specifically detects SHMT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
SHMT1 | |
Monoclonal | |
DyLight 755 | |
14 kDa protein, CSHMT, cytoplasmic serine hydroxymethyltransferase, Glycine hydroxymethyltransferase, MGC15229, MGC24556, serine hydroxymethyltransferase 1 (soluble), serine hydroxymethyltransferase, cytosolic, Serine methylase, SHMTEC 2.1.2.1 | |
Recombinant human SHMT1 (1-483aa) purified from E. coli | |
0.1 mL | |
metabolism | |
6470 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
AT26E5 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction