Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHMT1 Monoclonal Antibody (9C7)
GREENER_CHOICE

Catalog No. PIMA549246
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549246 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549246 Supplier Invitrogen™ Supplier No. MA549246
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: monkey liver tissue, rat liver tissue, mouse liver tissue. IHC: human liver cancer tissue. ICC/IF: A549 cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes the cellular form of serine hydroxymethyltransferase, a pyridoxal phosphate-containing enzyme that catalyzes the reversible conversion of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. This reaction provides one carbon units for synthesis of methionine, thymidylate, and purines in the cytoplasm. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing of this gene results in 2 transcript variants encoding 2 different isoforms. Additional transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHMT1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 9C7
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Shmt1
Gene Accession No. P34896, P50431
Gene Alias AI324848; AI385541; C81125; CSHMT; cytoplasmic serine hydroxymethyltransferase; cytosolic form; cytosolic serine hydroxymethyltransferase; glycine hydroxymethyltransferase; LRRGT00032; MEL-32; MGC15229; MGC24556; mshmt; mshmt1; mshmt2; serine hydroxymethyl transferase; serine hydroxymethyltransferase; serine hydroxymethyltransferase 1; serine hydroxymethyltransferase 1 (soluble); serine hydroxymethyltransferase 1 (soluble) isoform 1; serine hydroxymethyltransferase 2 (mitochondrial); serine hydroxymethyltransferase, cytosolic; Serine methylase; SHMT; Shmt1; shmt1 protein; SHMT2; zgc:66171; zgc:77524; zSHMT-1
Gene Symbols Shmt1
Host Species Mouse
Immunogen E.coli-derived human SHMT1 recombinant protein (Position: M1-S470).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20425, 287379, 6470
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.