Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527858
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527858 Supplier Invitrogen™ Supplier No. PA527858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (94%), Rat (95%), Zebrafish (83%), Xenopus laevis (84%), Rabbit (94%), Sheep (95%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the cellular form of serine hydroxymethyltransferase, a pyridoxal phosphate-containing enzyme that catalyzes the reversible conversion of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. This reaction provides one carbon units for synthesis of methionine, thymidylate, and purines in the cytoplasm. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing of this gene results in 2 transcript variants encoding 2 different isoforms. Additional transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHMT1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Shmt1
Gene Accession No. P34896
Gene Alias AI324848; AI385541; C81125; CSHMT; cytoplasmic serine hydroxymethyltransferase; cytosolic form; cytosolic serine hydroxymethyltransferase; glycine hydroxymethyltransferase; LRRGT00032; MEL-32; MGC15229; MGC24556; mshmt; mshmt1; mshmt2; serine hydroxymethyl transferase; serine hydroxymethyltransferase; serine hydroxymethyltransferase 1; serine hydroxymethyltransferase 1 (soluble); serine hydroxymethyltransferase 1 (soluble) isoform 1; serine hydroxymethyltransferase 2 (mitochondrial); serine hydroxymethyltransferase, cytosolic; Serine methylase; SHMT; Shmt1; shmt1 protein; SHMT2; zgc:66171; zgc:77524; zSHMT-1
Gene Symbols Shmt1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SHMT1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6470
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.