Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SHMT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543278
Description
Peptide sequence: NIIKKESNRQ RVGLELIASE NFASRAVLEA LGSCLNNKYS EGYPGQRYYG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Yeast: 100%; Zebrafish: 100%.
This gene encodes the cellular form of serine hydroxymethyltransferase, a pyridoxal phosphate-containing enzyme that catalyzes the reversible conversion of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. This reaction provides one carbon units for synthesis of methionine, thymidylate, and purines in the cytoplasm. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing of this gene results in 2 transcript variants encoding 2 different isoforms. Additional transcript variants have been described, but their biological validity has not been determined.
Specifications
| SHMT1 | |
| Polyclonal | |
| Unconjugated | |
| Shmt1 | |
| AI324848; AI385541; C81125; CSHMT; cytoplasmic serine hydroxymethyltransferase; cytosolic form; cytosolic serine hydroxymethyltransferase; glycine hydroxymethyltransferase; LRRGT00032; MEL-32; MGC15229; MGC24556; mshmt; mshmt1; mshmt2; serine hydroxymethyl transferase; serine hydroxymethyltransferase; serine hydroxymethyltransferase 1; serine hydroxymethyltransferase 1 (soluble); serine hydroxymethyltransferase 1 (soluble) isoform 1; serine hydroxymethyltransferase 2 (mitochondrial); serine hydroxymethyltransferase, cytosolic; Serine methylase; SHMT; Shmt1; shmt1 protein; SHMT2; zgc:66171; zgc:77524; zSHMT-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6470 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P34896 | |
| Shmt1 | |
| Synthetic peptide directed towards the N-terminal of human SHMT1 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction